missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rift Valley Fever Virus Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 178.00 - € 507.00
Specifications
| Antigen | Rift Valley Fever Virus |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613976
|
Novus Biologicals
NBP2-41082-0.025mg |
0.025 mg |
€ 178.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18110559
|
Novus Biologicals
NBP2-41082 |
0.1 mg |
€ 507.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rift Valley Fever Virus Polyclonal specifically detects Rift Valley Fever Virus in Virus samples. It is validated for ELISA.Specifications
| Rift Valley Fever Virus | |
| ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of the RVF virus. The immunogen is located within amino acids 960 - 1010 of Rift Valley Fever Virus. Amino Acid Squence: FERGSLPQTRNDKTFAASK | |
| Primary | |
| This RVF virus antibody was derived from a peptide sequence near the carboxy terminus of the polyprotein precursor translated from the M segment. It will therefore detect both the precursor and the Glycoprotein G2. |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Virus | |
| Glycoprotein, RVFV_sM_gp1 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title