missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rift Valley Fever Virus Polymerase Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 178.00 - € 507.00
Specifications
| Antigen | Rift Valley Fever Virus Polymerase |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18604399
|
Novus Biologicals
NBP2-41224-0.025mg |
0.025 mg |
€ 178.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18114388
|
Novus Biologicals
NBP2-41224 |
0.1 mg |
€ 507.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rift Valley Fever Virus Polymerase Polyclonal specifically detects Rift Valley Fever Virus Polymerase in Virus samples. It is validated for ELISA.Specifications
| Rift Valley Fever Virus Polymerase | |
| ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against a 16 amino acid peptide near the amino terminus of the RVF virus Polymerase. The immunogen is located within amino acids 370 - 420 of Rift Valley Fever Virus Polymerase. Amino Acid Squence: VKDRPDERNRYHRVHL | |
| Primary | |
| 230 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Virus | |
| EC=2.7.7.48, Replicase, RVFV_sL_gp1, Transcriptase | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title