missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rift Valley Fever Virus Polymerase Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41224F
This item is not returnable.
View return policy
Description
Rift Valley Fever Virus Polymerase Polyclonal antibody specifically detects Rift Valley Fever Virus Polymerase in Virus samples. It is validated for ELISA
Specifications
| Rift Valley Fever Virus Polymerase | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
| ELISA | |
| FITC | |
| EC=2.7.7.48, Replicase, RVFV_sL_gp1, Transcriptase | |
| Antibody was raised against a 16 amino acid peptide near the amino terminus of the RVF virus Polymerase. The immunogen is located within amino acids 370 - 420 of Rift Valley Fever Virus Polymerase. Amino Acid Squence: VKDRPDERNRYHRVHL | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 9538294 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction