missing translation for 'onlineSavingsMsg'
Learn More

RIG-I Antibody (4G1B6), Novus Biologicals™

Product Code. p-200080643 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18051504 0.1 mL 0.1mL
18654747 0.025 mL 0.03mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18051504 Supplier Novus Biologicals Supplier No. NBP261849

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 1 publication

RIG-I Monoclonal specifically detects RIG-I in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIG-I
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 4G1B6
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000
Formulation PBS with 0.05% Sodium Azide
Gene Alias DEAD (Asp-Glu-Ala-Asp) box polypeptide 58, DEAD box protein 58, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, DKFZp686N19181, EC 3.6.4.13, FLJ13599, probable ATP-dependent RNA helicase DDX58, retinoic acid inducible gene I, Retinoic acid-inducible gene 1 protein, Retinoic acid-inducible gene I protein, RIG-1, RIG-IDKFZp434J1111, RNA helicase RIG-I
Gene Symbols RIGI
Host Species Mouse
Immunogen Purified recombinant fragment of human DDX58 (AA: 789-925) expressed in E. Coli.
Molecular Weight of Antigen 106.6 kDa
Purification Method Size Exclusion Chromatography
Quantity 0.1 mL
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 23586
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.