missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RIG-I Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142289
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
Retinoic acid-inducible gene I, RIG-I is a pattern recognition receptor (PRR) involved in the recognition of viral dsRNA. Along with MDA5, RIG-I detects viral dsRNA and activates the innate immune response. Both MDA5 and RIG-I are RNA helicases and they perform overlapping as well as distinct roles. RIG-I is activated by dsRNAs without a 5'-triphosphate end and short dsRNAs, whereas MDA5 is activated by long dsRNAs. Once activated, both proteins signal through IPS-1 activating transcription factors NF-kappaB and IRF-3 (1) and ultimately activating apoptosis, cytokine signaling, and inflammation. RIG-I is essential for signaling by influenza A, influenza B, human respiratory syncytial virus (3), paromyxoviruses, Japanese encephalitis virus, and West Nile virus. MicroRNA-146a has been implicated in feedback inhibition of RIG-I-dependant antiviral response by negatively regulating RIG-I targets TRAF6, IRAK1, and IRAK2. Recent evidence has implicated RIG-I in the detection of cytosolic DNA through RNA polymerase III activity.
Specifications
| RIG-I | |
| Polyclonal | |
| Unconjugated | |
| DDX58 | |
| 6430573D20Rik; C330021E21; Ddx58; DEAD (Asp-Glu-Ala-Asp) box polypeptide 58; DEAD box protein 58; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide; DEAD/H box polypeptide RIG-I; DEAD-box protein 58; DEXD/H-box helicase 58; probable ATP-dependent RNA helicase DDX58; Retinoic acid-inducible gene 1 protein; retinoic acid-inducible gene I protein; retinoic acid-inducible gene-I; RIG-1; RIGI; RIG-I; RIG-I-like receptor 1; RLR-1; RNA helicase RIG-I; SGMRT2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 23586 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O95786 | |
| DDX58 | |
| Peptide with sequence C-KDFHFEKIPFDPAE. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction