missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rigosertib, MedChemExpress
Description
Rigosertib (ON-01910) is a multi-kinase inhibitor and a selective anti-cancer agent, which induces apoptosis by inhibition the PI3 kinase/Akt pathway, promots the phosphorylation of histone H2AX and induces G2/M arrest in cell cycle. Rigosertib is a selective and non-ATP-competitive inhibitor of PLK1 with an IC50 of 9 nM.
Specifications
Specifications
| Chemical Name or Material | Rigosertib |
| CAS | 592542-59-1 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C21H25NO8S |
| Quantity | 100 mg |
| Synonym | ON-01910 |
| Solubility Information | DMSO : 75 mg/mL (166.12 mM; Need ultrasonic) |
| SMILES | COC1=CC=C(C=C1NCC(O)=O)CS(/C=C/C2=C(C=C(C=C2OC)OC)OC)(=O)=O.[(E)] |
| Molecular Weight (g/mol) | 451.49 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?