Learn More
Riley™ Riley Tilla Safety Glasses
Click to view available options
Lens Tint:
Clear
Gray
Unit Size:
Pack of 20
Description
- Can be worn over spectacles
- Ventilated temple tips and side shields
- Brow protection
- Anti-fog lens
- UV400 protection
- Ideal to be worn by site visitors
Specifications
Specifications
| Certifications/Compliance | CE or - UKCA or - EN 166:2001 |
| Lens Coating | Moisture Repellent, UV-treated |
| Lens Tint | Clear |
| Product Type | Overglasses |
| For Use With (Application) | Component and Product Assembly, Woodwork and Fabrication, Glass and Glazing, Surveying |
| Frame Color | Clear |
| Transmittance Value | 92% |
| Weight (Metric) | 44 g |
| Material | Polycarbonate |
| Disposable | Reusable |
| Show More |