missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rilvegostomig Recombinant Human Monoclonal Antibody

Rilvegostomig Recombinant Monoclonal Antibody
€ 610.00 - € 2530.00
Specifications
| Antigen | Rilvegostomig |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30284737
![]() |
Invitrogen
MA560159 |
100 μg |
€ 610.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30284109
|
Invitrogen
MA560160 |
1 mg |
€ 2530.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rilvegostomig Recombinant Monoclonal Antibody for SPR, Flow, ELISA, Functional
Specifications
| Rilvegostomig | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Lyophilized | |
| IgG1 | |
| Human | |
| Primary | |
| Protein A |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| Unconjugated | |
| Human | |
| RUO | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title