missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIMS2 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76905B
This item is not returnable.
View return policy
Description
RIMS2 Polyclonal antibody specifically detects RIMS2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| RIMS2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Biotin | |
| DKFZp781A0653, KIAA0751RIM2OBOERAB3 interacting protein 3, non-small cell lung cancer RimL3a protein, non-small cell lung cancer RimL3c protein, nuclear protein, rab3-interacting molecule 2, Rab-3-interacting molecule 2, Rab-3-interacting protein 3, RAB3IP3, regulating synaptic membrane exocytosis 2, regulating synaptic membrane exocytosis protein 2, RIM 2 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the center of the human Rim2. The immunogen is located within amino acids 250 - 300 of Rim2. Amino Acid Squence: DRKRSPSVSRDQNRRYD | |
| 0.1 mL | |
| Neuroscience | |
| 9699 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction