missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RIN3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558949
This item is not returnable.
View return policy
Description
Immunogen sequence: AKKNLPTAPP RRRVSERVSL EDQSPGMAAE GDQLSLPPQG TSDGPEDTPR ESTEQGQDTE VKASDPHSMP ELPRTAKQPP VPPPRKKRIS RQLASTLPA Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 58%.
RIN3 is a member of the RIN family of Ras interaction-interference proteins, which are binding partners to the RAB5 small GTPases (Kajiho et al., 2003 ).
Specifications
| RIN3 | |
| Polyclonal | |
| Unconjugated | |
| RIN3 | |
| 6430500K07; C86933; RAB5 interacting protein 3; ras and Rab interactor 3; Ras interaction/interference protein 3; Rin3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79890 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TB24 | |
| RIN3 | |
| Recombinant protein corresponding to Human RIN3. Recombinant protein control fragment (Product #RP-96512). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction