missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ring finger protein 138 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 193.00 - € 470.00
Specifications
| Antigen | Ring finger protein 138 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18643580
|
Novus Biologicals
NBP2-93425-0.02ml |
0.02 mL |
€ 193.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18698270
|
Novus Biologicals
NBP2-93425-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ring finger protein 138 Polyclonal antibody specifically detects Ring finger protein 138 in Human samples. It is validated for Western BlotSpecifications
| Ring finger protein 138 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Zinc Finger | |
| PBS (pH 7.3), 50% glycerol | |
| 51444 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| E3 ubiquitin-protein ligase RNF138, EC 6.3.2.-, hNARF, HSD-4, MGC8758, NARF, Nemo-like kinase-associated RING finger protein, ring finger protein 138NLK-associated RING finger protein, STRIN | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-245 of mouse Ring finger protein 138 (Q9CQE0). SVTRRERACPERALDLENIMRRFSGSCRCCSKKIKFYRMRHHYKSCKKYQDEYGVSSVIPNFKISQDSVRSSNRSETSASDNTETYQEDTSSSGHPTFKCPLCQESNFTRQRLLDHCNSNHLFQIVPVTCPICVSLPWGDPSQITRNFVSHLNQRHQFDYGEFVNLQLDEETQYQTAVEESFQVNM | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title