Learn More
Emma Safety Footwear Ringo All-Round Protection Versatile D-Fit Shoes
Designed with wipe-clean uppers and easy-clean sole.
Brand: Emma Safety Footwear MM73054038
Additional Details : Weight : 0.01000kg
Description
- Full grain leather uppers
- Hydro-Tec™ sanitized silver lining
- Non-woven antistatic insole
- GripForce™ Easy Twist outsole
Specifications
Hydro-Tec Sanitized Silver lining | |
Non-woven antistatic insole | |
Steel Toe |
38 | |
GripForce™ Easy Twist outsole | |
All-Round Protection Versatile Shoes |