missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RINT1 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00060561-B01P
This item is not returnable.
View return policy
Description
RINT1 Polyclonal antibody specifically detects RINT1 in Human samples. It is validated for Western Blot
Specifications
| RINT1 | |
| Polyclonal | |
| Western Blot 1:500 | |
| NP_068749.3 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| DKFZp667H2324, FLJ11785, HsRINT-1, RAD50 interactor 1hsRINT-1, RINT-1RAD50-interacting protein 1 | |
| RINT-1 (NP_068749.3, 1 a.a. - 792 a.a.) full-length human protein. MLPAGEIGASPAAPCCSESGDERKNLEEKSDINVTVLIGSKQVSEGTDNGDLPSYVSAFIEKEVGNDLKSLKKLDKLIEQRTVSKMQLEEQVLTISSEIPKRIRSALKNAEESKQFLNQFLEQETHLFSAINSHLLTAQPWMDDLGTMISQIEEIERHLAYLKWISQIEELSDNIQQYLMTNNVPEAASTLVSMAELDIKLQESSCTHLLGFMRATVKFWHKILKDKLTSDFEEILAQLHWPFIAPPQSQTVGLSRPASAPEIYSYLETLFCQLLKLQTSDELLTEPKQLPEKYSLPASPSVILPIQVMLTPLQKRFRYHFRGNRQTNVLSKPEWYLAQVLMWIGNHTEFLDEKIQPILDKVGSLVNARLEFSRGLMMLVLEKLATDIPCLLYDDNLFCHLVDEVLLFERELHSVHGYPGTFASCMHILSEETCFQRWLTVERKFALQKMDSMLSSEAAWVSQYKDITDVDEMKVPDCAETFMTLLLVITDRYKNLPTASRKLQFLELQKDLVDDFRIRLTQVMKEETRASLGFRYCAILNAVNYISTVLADWADNVFFLQLQQAALEVFAENNTLSKLQLGQLASMESSVFDDMINLLERLKHDMLTRQVDHVFREVKDAAKLYKKERWLSLPSQSEQAVMSLSSSACPLLLTLRDHLLQLEQQLCFSLFKIFWQMLVEKLDVYIYQEIILANHFNEGGAAQLQFDMTRNLFPLFSHYCKRPENYFKHIKEACIVLNLNVGSALLLKDVLQSASGQLPATAALNEVGIYKLAQQDVEILLNLRTNWPNTGK | |
| 0.05 mg | |
| Cell Cycle and Replication | |
| 60561 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction