Learn More
RIOK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17222-1-AP-20UL
This item is not returnable.
View return policy
Description
FXR1 is an RNA binding protein that interacts with the functionally-similar proteins FMR1 and FXR2. It may regulate intracellular transport and local transltion of certain mRNAs. These proteins shuttle between the nucleus and cytoplasm and associate with polyribosomes, predomitly with the 60S ribosomal subunit. FXR1 is required for embryonic and postnatal development of muscle tissue. Three transcript variants encoding different isoforms have been found for this gene.Specifications
| RIOK1 | |
| Polyclonal | |
| Unconjugated | |
| RIOK1 | |
| AD034, RIO kinase 1, RIO kinase 1 (yeast), RIOK1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 291061, 71340, 83732 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q922Q2, Q9BRS2 | |
| Riok1 | |
| RIOK1 Fusion Protein Ag11057 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |