missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIOK2 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-30099H
This item is not returnable.
View return policy
Description
RIOK2 Polyclonal antibody specifically detects RIOK2 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| RIOK2 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| EC 2.7.11.1, FLJ11159, RIO kinase 2, RIO kinase 2 (yeast), RIO2, serine/threonine-protein kinase RIO2 | |
| Antigen maps to a region between residue 502 and 552 of human RIO kinase 2 using the numbering given in entry NP_060813.1 | |
| 0.1 mL | |
| Protein Kinase | |
| 55781 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction