missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RIOK3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16517931
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16517931 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16517931 Supplier Invitrogen™ Supplier No. PA590666

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: NHVFNALKQH AYSEERRSAR LHEKKEHSTA EKAVDPKTRL LMYKMVNSGM LETITGCIST GKESVVFHAY GGSMEDEKED SKVIPTECAI KVFKTTLNEF KNRDKYIKDD FRFKDRFSKL NPRKIIRMWA EKEMHNLARM QRAGIPCPTV VLLKKHILVM SFIGHDQVPA PKLKEVKLNS EEMKEAYYQT LHLMRQLYHE CTLVHADLSE YNMLWHAGKV WLIDVSQSVE PTHPHGLEFL FRDCRNVSQF FQKGGVKEAL SERELFNAVS GLNITADNEA DFLAEIEALE KMNEDHVQKN GRKAASFLKD DGDPPLLYDE; Positive Samples: U-251MG, Mouse pancreas.

The phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions in eukaryotes, including cell division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the serine/threonine (Ser/Thr) protein kinases. SUDD, also known as RIOK3 (RIO kinase 3), is a 519 amino acid protein that contains one protein kinase domain and belongs to the Ser/Thr protein kinase family. Expressed in a variety of tissues, SUDD catayzes the ATP-dependent phosphorylation of target proteins, thereby influencing signaling events throughout the cell. SUDD is expressed as two isoforms due to alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIOK3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RIOK3
Gene Accession No. O14730, Q9DBU3
Gene Alias 1200013N13Rik; D18Ertd331e; E130306C24Rik; homolog of the Aspergillus nidulans sudD gene product; RIO kinase 3; RIOK3; Serine/threonine-protein kinase RIO3; SUDD; sudD (suppressor of bimD6, Aspergillus nidulans) homolog; sudD homolog; sudD suppressor of Aspergillus nidulans bimD6 homolog; sudD suppressor of bimD6 homolog; sudD, suppressor of bimD6 homolog; testicular secretory protein Li 47
Gene Symbols RIOK3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 200-519 of human RIOK3 (NP_0038222).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66878, 8780
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.