missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIP5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12785-100ul
This item is not returnable.
View return policy
Description
RIP5 Polyclonal antibody specifically detects RIP5 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| RIP5 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:100-1:500 | |
| C20orf24, chromosome 20 open reading frame 24, PNAS-11 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human RIP5 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 55969 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction