Learn More
RIPA-56, MedChemExpress
Product Code. 16490916
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490916 200 mg 200mg
16459786 50 mg 50mg
16444506 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16490916 Supplier Medchem Express Supplier No. HY101032
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress RIPA-56 is a highly potent, selective, and metabolically stable inhibitor of receptor-interacting protein 1 (RIP1) with an IC50 of 13 nM. RIPA-56 can be used for the treatment of systemic inflammatory response syndrome.
RIPA-56 is a highly potent, selective, and metabolically stable inhibitor of receptor-interacting
Specifications
Chemical Name or Material RIPA-56
Physical Form Solid
CAS 1956370-21-0
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C13H19NO2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 100 mg/mL (451.88 mM)
SMILES CCC(C)(C)C(N(CC1=CC=CC=C1)O)=O
Molecular Weight (g/mol) 221.3
Formula Weight 221.3
Percent Purity 99.71%
Grade Research
Show More Show Less
Research purposes only