missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIPK1 Monoclonal Antibody (OTI2F4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800311S
This item is not returnable.
View return policy
Description
RIPK1 Monoclonal antibody specifically detects RIPK1 in Human samples. It is validated for Western BlotSpecifications
| RIPK1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13546 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8737 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2F4 | |
| Unconjugated | |
| RIPK1 | |
| cell death protein RIP; D330015H01Rik; FLJ39204; OTTHUMP00000015955; receptor (TNFRSF)-interacting serine-threonine kinase 1; receptor interacting serine/threonine kinase 1; receptor-interacting protein 1; receptor-interacting protein kinase 1; receptor-interacting serine/threonine-protein kinase 1; Rinp; RIP; RIP1; RIP-1; RIP1 (CT); RIPK1; serine/threonine-protein kinase RIP | |
| Human recombinant protein fragment corresponding to amino acids 133-422 of human RIPK1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction