Learn More
RIPK1/RIP1 Antibody (CL11368) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-15118AF532
Product Code. 30288871
  • € 737.00 / 0.10mL
Description
RIPK1/RIP1 Monoclonal antibody specifically detects RIPK1/RIP1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RIPK1/RIP1
Monoclonal
Alexa Fluor 532
Cell death protein RIP, EC 2.7.11.1, receptor (TNFRSF)-interacting serine-threonine kinase 1, receptor interacting protein, Receptor-interacting protein 1, receptor-interacting serine/threonine-protein kinase 1, RIP1, RIP-1, Rip1 kinase, RIPFLJ39204, Serine/threonine-protein kinase RIP
This antibody was developed using a recombinant protein derived from Q13546, with the exact immunogen sequence remaining proprietary.
0.1 mL
Apoptosis, Necroptosis
8737
Store at 4⪚C in the dark.
IgG1
Western Blot
CL11368
50mM Sodium Borate
Mouse
Protein A purified