missing translation for 'onlineSavingsMsg'
Learn More

RIPK1/RIP1 Antibody (CL11368) [Alexa Fluor« 750], Novus Biologicals Biologicals™

Product Code. 30528243 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30528243 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30528243 Supplier Novus Biologicals Supplier No. NBP344182AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RIPK1/RIP1 Monoclonal antibody specifically detects RIPK1/RIP1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIPK1/RIP1
Applications Western Blot
Classification Monoclonal
Clone CL11368
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias Cell death protein RIP, EC 2.7.11.1, receptor (TNFRSF)-interacting serine-threonine kinase 1, receptor interacting protein, Receptor-interacting protein 1, receptor-interacting serine/threonine-protein kinase 1, RIP1, RIP-1, Rip1 kinase, RIPFLJ39204, Serine/threonine-protein kinase RIP
Host Species Mouse
Immunogen This antibody was generated using a recombinant protein sequence of Q13546, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Necroptosis
Primary or Secondary Primary
Gene ID (Entrez) 8737
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.