missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIPK3 Monoclonal Antibody (OTI2F11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803095S
This item is not returnable.
View return policy
Description
RIPK3 Monoclonal antibody specifically detects RIPK3 in Human samples. It is validated for Western BlotSpecifications
| RIPK3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y572 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11035 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI2F11 | |
| Unconjugated | |
| RIPK3 | |
| 2610528K09Rik; AW107945; Hcyp2; Homocysteine respondent protein HCYP2; mRIP3; receptor interacting protein 3; receptor interacting serine/threonine kinase 3; receptor-interacting protein 3; receptor-interacting serine/threonine-protein kinase 3; receptor-interacting serine-threonine kinase 3; Rip3; RIP-3; Ripk3; RIP-like protein kinase 3 | |
| Human recombinant protein fragment corresponding to amino acids 233-518 of human RIPK3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction