Learn More
RIPK3 Monoclonal Antibody (OTI3A12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA803170S
Encompass_Preferred
 View more versions of this product
Product Code. 17582093
  • € 242.00 / 30µL
Description
RIPK3 Monoclonal antibody specifically detects RIPK3 in Human samples. It is validated for Western Blot
Specifications
Specifications
RIPK3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O00469
Mouse
Affinity chromatography
RUO
5352
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI3A12
Unconjugated
Plod2
2610528K09Rik; AW107945; Hcyp2; Homocysteine respondent protein HCYP2; mRIP3; receptor interacting protein 3; receptor interacting serine/threonine kinase 3; receptor-interacting protein 3; receptor-interacting serine/threonine-protein kinase 3; receptor-interacting serine-threonine kinase 3; Rip3; RIP-3; Ripk3; RIP-like protein kinase 3
Human recombinant protein fragment corresponding to amino acids 359-640 of human PLOD2 produced in E.coli.