missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIPK3/RIP3 Antibody (1312A), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-76800AF532
This item is not returnable.
View return policy
Description
RIPK3/RIP3 Monoclonal antibody specifically detects RIPK3/RIP3 in Human samples. It is validated for Western Blot
Specifications
| RIPK3/RIP3 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3 | |
| RIPK3/RIP3 Antibody (1312A) [Alexa Fluor 532] was made to a partial synthetic peptide made to an internal portion of the human RIPK3/RIP3 protein (between amino acids 470-540). This seqeunce has 100% homology to isoform CRA_a and isoform CRA_b. | |
| 0.1 mL | |
| Cancer, Cell Biology, Immunology, Necroptosis, Protein Kinase, Signal Transduction | |
| 11035 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| 1312A | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction