missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RIPK4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16519921
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16519921 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16519921 Supplier Invitrogen™ Supplier No. PA597085

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: HRPELPPVCR ARPRACSHLI RLMQRCWQGD PRVRPTFQEI TSETEDLCEK PDDEVKETAH DLDVKSPPEP RSEVVPARLK RASAPTFDND YSLSELLSQL DSGVSQAVEG PEELSRSSSE SKLPSSGSGK RLSGVSSVDS AFSSRGSLSL SFEREPSTSD LGTTDVQKKK LVDAIVSGDT SKLMKILQPQ DVDLALDSGA SLLHLAVEAG QEECAKWLLL NNANPNLSNR RGSTPLHMAV ERRVRGVVEL LLARKISVNA KDEDQWTALH FAAQNGDESS T; Positive Samples: 293T, Mouse liver; Cellular Location: Cytoplasm, Membrane, Peripheral membrane protein.

The protein encoded by this gene is a serine/threonine protein kinase that interacts with protein kinase C-delta. The encoded protein can also activate NFkappaB and is required for keratinocyte differentiation. This kinase undergoes autophosphorylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIPK4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RIPK4
Gene Accession No. P57078, Q9ERK0
Gene Alias 2310069J12Rik; AI552420; ANKK2; ANKRD3; ankyrin repeat domain 3; ankyrin repeat domain-containing protein 3; DIK; NKRD3; PKC-associated protein kinase; PKC-delta-interacting protein kinase; PKC-regulated protein kinase; pkk; PPS2; protein kinase C-associated kinase; protein kinase PKK; receptor interacting serine/threonine kinase 4; receptor-interacting serine/threonine-protein kinase 4; receptor-interacting serine-threonine kinase 4; RIP4; Ripk4; serine/threonine-protein kinase ANKRD3; wu:fj80b10; zgc:55705
Gene Symbols RIPK4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 240-520 of human RIPK4 (NP_0656902).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54101, 72388
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.