missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIPPLY2 Antibody (OTI1B5), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73913AF647
This item is not returnable.
View return policy
Description
RIPPLY2 Monoclonal specifically detects RIPPLY2 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| RIPPLY2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RIPPLY2 | |
| Full length recombinant protein of human RIPPLY2 (NP_001009994) produced in HEK293T cells. | |
| 0.1 mL | |
| 134701.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| OTI1B5 | |
| Western Blot, Flow Cytometry | |
| C6orf159, C6orf59, Chromosome 6 Open Reading Frame 159, Ripply Transcriptional Repressor 2, Ripply2 Homolog, Ripply2 Homolog (Zebrafish) | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction