Learn More
Invitrogen™ Risankizumab Humanized Recombinant Human Monoclonal Antibody
GREENER_CHOICE
Human Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA542076
Product Code. 17788693
  • € 502.00 / 100µg
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Risankizumab is a fully humanized IgG1 monoclonal antibody (mAb) that is selective for interleukin 23 (IL-23). On April 23, 2019 it was approved by the FDA (as SKYRIZI) for the treatment of moderate-to-severe plaque psoriasis in adults who are clinically considered eligible for systemic therapy or phototherapy for psoriasis. Label This pharmaceutical has been commercialized through collaboration between the German pharmaceutical company, Boehringer Ingelheim, and Abbvie.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Risankizumab Humanized
Recombinant Monoclonal
Unconjugated
BI 655066; risankizumab-rzaa
Human IL23A.
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Chemical
ELISA, SDS-Page
1 mg/mL
PBS with no preservative; pH 7.4
Human
Protein A/G