Learn More
Invitrogen™ Risankizumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559565
 View more versions of this product
Product Code. 30282651
  • € 477.00 / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Risankizumab is a fully humanized IgG1 monoclonal antibody (mAb) that is selective for interleukin 23 (IL-23). On April 23, 2019 it was approved by the FDA (as SKYRIZI) for the treatment of moderate-to-severe plaque psoriasis in adults who are clinically considered eligible for systemic therapy or phototherapy for psoriasis. Label This pharmaceutical has been commercialized through collaboration between the German pharmaceutical company, Boehringer Ingelheim, and Abbvie.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Risankizumab Humanized
Recombinant Monoclonal
Unconjugated
BI 655066; risankizumab-rzaa
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized