missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RISC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA129824
This item is not returnable.
View return policy
Description
A suggested positive control is tested with human heart cell lysate.
The RNA-induced silencing complex (RISC) is a key player in the regulation of gene expression through RNA interference (RNAi) mechanisms. It operates by utilizing small RNA molecules-such as microRNAs (miRNAs), small interfering RNAs (siRNAs), and PIWI-interacting RNAs (piRNAs)-which are processed from double-stranded RNA precursors by enzymes like Drosha and Dicer in animals, and DICER-LIKE in plants. RISC is a versatile molecular complex consisting of Argonaute proteins as its core component, which guides the complex to the target RNA by complementary base pairing. Once assembled, RISC can recruit other proteins that mediate gene silencing by degrading or inhibiting the translation of the target RNA. This post-transcriptional regulation system is integral to numerous biological processes, allowing cells to control gene expression finely and respond to endogenous and exogenous signals. The functionality of RISC in gene silencing highlights its significance in the broader context of genetic regulation and has implications in therapies that target gene expression through RNAi.
Specifications
| RISC | |
| Polyclonal | |
| Unconjugated | |
| SCPEP1 | |
| 2410018F01Rik; 4833411K15Rik; AI957256; HSCP1; MSTP034; retinoid inducible serine carboxypeptidase; retinoid-inducible serine carboxypeptidase; retinoid-inducible serine caroboxypeptidase; retinoid-inducible serine caroboxypetidase; Risc; SCP1; SCPEP1; Serine carboxypeptidase 1; serine carboxypeptidase 1 precursor protein; serine caroboxypeptidase 1; UNQ265/PRO302 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 114861, 59342, 74617 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q920A5, Q920A6, Q9HB40 | |
| SCPEP1 | |
| Synthetic peptide corresponding to aa 28-43 of human RISC. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction