missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rit2 Antibody (OTI4A6), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73914AF700
This item is not returnable.
View return policy
Description
Rit2 Monoclonal specifically detects Rit2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Rit2 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RIT2 | |
| Full length recombinant protein of human Rit2 (NP_002921) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot | |
| OTI4A6 | |
| Western Blot | |
| GTP-binding protein Rit2, GTP-binding protein Roc2, Ras-like protein expressed in neurons, Ras-like without CAAX 2, Ras-like without CAAX protein 2, RIBA, Ric (Drosophila)-like, expressed in neurons, Ric-like, expressed in neurons, RIN, ROC2 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 6014.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction