missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIT2 Monoclonal Antibody (OTI4B5), TrueMAB™, OriGene
Description
RIT2 Monoclonal antibody specifically detects RIT2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications
Specifications
| Antigen | RIT2 |
| Applications | Flow Cytometry, Immunocytochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI4B5 |
| Concentration | 0.84 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | RIT2 |
| Gene Accession No. | Q99578 |
| Gene Alias | GTP-binding protein Rit2; GTP-binding protein Roc2; Ras like without CAAX 2; RAS-like protein expressed in neuron; ras-like protein expressed in neurons; Ras-like without CAAX 1; Ras-like without CAAX 2; ras-like without CAAX protein 2; RIBA; Ric-like, expressed in neurons; RIN; Rit1; RIT2; ROC2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?