Learn More
Rituximab Mouse, FITC, Monoclonal Antibody, AcroBiosystems
Product Code. 17136651
Change view
Click to view available options
Quantity:
200 μg
25 μg
Unit Size:
200µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17136651 25 μg 25µg
17126651 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17136651 Supplier ACROBiosystems Supplier No. RIBFY35c25ug
Mouse Monoclonal Antibody
Rituximab Monoclonal antibody specifically detects Rituximab.
Specifications
Antigen Rituximab
Classification Monoclonal
Conjugate FITC
Host Species Hybridoma
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C, aviod light
Isotype IgG1