Learn More
Rituximab Mouse, Unconjugated, Monoclonal Antibody, AcroBiosystems
Description
Rituximab Monoclonal antibody specifically detects Rituximab.
Specifications
Specifications
| Antigen | Rituximab |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Hybridoma |
| Quantity | 100 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Content And Storage | -20°C |