Learn More
Rituximab Mouse, Unconjugated, Monoclonal Antibody, AcroBiosystems
Product Code. 16418019
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16418019 1 mg 1mg
16408019 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16418019 Supplier ACROBiosystems Supplier No. RIBY371mg
Mouse Monoclonal Antibody
Rituximab Monoclonal antibody specifically detects Rituximab.
Specifications
Antigen Rituximab
Classification Monoclonal
Conjugate Unconjugated
Host Species Hybridoma
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C