missing translation for 'onlineSavingsMsg'
Learn More

Rivaroxaban Pseudodimer, TRC

Product Code. 30138212
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138212 50 mg 50mg
30133167 25 mg 25mg
30008151 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138212 Supplier Toronto Research Chemicals Supplier No. TRCR53801550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1632463-24-1
Molecular Formula C38 H36 Cl2 N6 O10 S2
Molecular Weight (g/mol) 871.76
Synonym 2-Thiophenecarboxamide, 5-chloro-N-[4-[(5S)-5-[[[(5-chloro-2-thienyl)carbonyl]amino]methyl]-2-oxo-3-oxazolidinyl]phenyl]-N-[2-[2-oxo-2-[[[(5S)-2-oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl]methyl]amino]ethoxy]ethyl]-, Rivaroxaban Imp. J;
IUPAC Name 5-chloro-N-[[(5S)-3-[4-[(5-chlorothiophene-2-carbonyl)-[2-[2-oxo-2-[[(5S)-2-oxo-3-[4-(3-oxomorpholin-4-yl)phenyl]-1,3-oxazolidin-5-yl]methylamino]ethoxy]ethyl]amino]phenyl]-2-oxo-1,3-oxazolidin-5-yl]methyl]thiophene-2-carboxamide
SMILES Clc1ccc(s1)C(=O)NC[C@H]2CN(C(=O)O2)c3ccc(cc3)N(CCOCC(=O)NC[C@H]4CN(C(=O)O4)c5ccc(cc5)N6CCOCC6=O)C(=O)c7ccc(Cl)s7

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C38H36Cl2N6O10S2/c39-31-11-9-29(57-31)35(49)42-18-28-20-46(38(52)56-28)26-7-3-24(4-8-26)44(36(50)30-10-12-32(40)58-30)14-16-53-21-33(47)41-17-27-19-45(37(51)55-27)25-5-1-23(2-6-25)43-13-15-54-22-34(43)48/h1-12,27-28H,13-22H2,(H,41,47)(H,42,49)/t27-,28-/m0/s1
Formula Weight 870.1311
Percent Purity >95%
Chemical Name or Material Rivaroxaban Pseudodimer
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.