missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIZ1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94500-0.02ml
This item is not returnable.
View return policy
Description
RIZ1 Polyclonal antibody specifically detects RIZ1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RIZ1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 2.1.1.43, GATA-3 binding protein G3B, GATA-3-binding protein G3B, KMT8PR domain-containing protein 2, Lysine N-methyltransferase 8, MTB-ZFHUMHOXY1, PR domain containing 2, with ZNF domain, PR domain zinc finger protein 2, retinoblastoma protein-binding zinc finger protein, Retinoblastoma protein-interacting zinc finger protein, RIZ1, RIZ2, RIZMTE-binding protein, Zinc finger protein RIZ, zinc-finger DNA-binding protein | |
| A synthetic peptide corresponding to a sequence within amino acids 1-176 of human RIZ1 (NP_036363.2). MNQNTTEPVAATETLAEVPEHVLRGLPEEVRLFPSAVDKTRIGVWATKPILKGKKFGPFVGDKKKRSQVKNNVYMWEVYYPNLGWMCIDATDPEKGNWLRYVNWACSGEEQNLFPLEINRAIYYKTLKPIAPGEELLVWYNGEDNPEIAAAIEEERASARSKRSSPKSRKGKKKSQ | |
| 0.02 mL | |
| Breast Cancer, Chromatin Research | |
| 7799 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction