missing translation for 'onlineSavingsMsg'
Learn More

RK-24466, MedChemExpress

Product Code. 16426897
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16426897

Brand: Medchem Express HY108318

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RK-24466 (KIN 001-51) is a potent and selective Lck inhibitor; inhibits Lck (64-509) and LckCD isoforms with IC50s of less than 1 and 2 nM, respectively.

RK-24466 (KIN 001-51) is a potent and selective Lck inhibitor

Specifications

Chemical Name or Material RK-24466
CAS 213743-31-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H22N4O
Quantity 5 mg
Synonym KIN 001-51
Solubility Information DMSO : 45 mg/mL (121.47 mM; Need ultrasonic)
SMILES NC1=C2C(N(C3CCCC3)C=C2C4=CC=C(OC5=CC=CC=C5)C=C4)=NC=N1
Molecular Weight (g/mol) 370.45
Formula Weight 370.45
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.