missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RKHD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA520622
This item is not returnable.
View return policy
Description
A suggested positive control is rat heart tissue lysate. PA5-20622 can be used with blocking peptide PEP-0742.
Rkhd2, also known as MEX3C is a member of a novel family of four homologous human MEX3 proteins each containing two heterogeneous nuclear ribonucleoprotein K homology (KH) domains and one carboxy-terminal RING finger module. MEX3 proteins, including Rkhd2, are phosphoproteins that bind RNA through their KH domains and shuttle between the nucleus and the cytoplasm via the CRM1 export pathway. These proteins are a novel family of evolutionarily conserved RNA-binding proteins, differentially recruited to P bodies and potentially involved in post-transcriptional regulatory mechanisms. It has been suggested that genetic variations in Rkhd2 may be associated with susceptibility to essential hypertension type 8. Rkhd3 and Rkhd4, but not Rkhd2, co-localize with both the hDcp1a decapping factor and Argonaute (Ago) proteins in processing bodies (P bodies), recently characterized as centers of mRNA turnover.
Specifications
| RKHD2 | |
| Polyclonal | |
| Unconjugated | |
| MEX3C | |
| A130001D14Rik; BC035207; BM-013; mex-3 homolog C; mex3 homolog C (C. elegans); mex-3 RNA binding family member C; Mex3c; MEX-3C; MEX3C variant 1; MEX3C variant 10; MEX3C variant 11; MEX3C variant 2; MEX3C variant 3; MEX3C variant 4; MEX3C variant 5; MEX3C variant 6; MEX3C variant 9; ring finger and KH domain containing 2; RING finger and KH domain-containing protein 2; RING finger protein 194; RING-type E3 ubiquitin transferase MEX3C; Rkhd2; RNA-binding E3 ubiquitin-protein ligase MEX3C; RNA-binding protein MEX3C; RNF194 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 240396, 307271, 51320 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q05A36, Q5U5Q3 | |
| MEX3C | |
| A 15 amino acid peptide from near the carboxy terminus of human RKHD2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction