missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RKHD2 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5142905

Product Code. 17858242

  • € 474.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.

Rkhd2, also known as MEX3C is a member of a novel family of four homologous human MEX3 proteins each containing two heterogeneous nuclear ribonucleoprotein K homology (KH) domains and one carboxy-terminal RING finger module. MEX3 proteins, including Rkhd2, are phosphoproteins that bind RNA through their KH domains and shuttle between the nucleus and the cytoplasm via the CRM1 export pathway. These proteins are a novel family of evolutionarily conserved RNA-binding proteins, differentially recruited to P bodies and potentially involved in post-transcriptional regulatory mechanisms. It has been suggested that genetic variations in Rkhd2 may be associated with susceptibility to essential hypertension type 8. Rkhd3 and Rkhd4, but not Rkhd2, co-localize with both the hDcp1a decapping factor and Argonaute (Ago) proteins in processing bodies (P bodies), recently characterized as centers of mRNA turnover.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RKHD2
Polyclonal
Unconjugated
MEX3C
A130001D14Rik; BC035207; BM-013; mex-3 homolog C; mex3 homolog C (C. elegans); mex-3 RNA binding family member C; Mex3c; MEX-3C; MEX3C variant 1; MEX3C variant 10; MEX3C variant 11; MEX3C variant 2; MEX3C variant 3; MEX3C variant 4; MEX3C variant 5; MEX3C variant 6; MEX3C variant 9; ring finger and KH domain containing 2; RING finger and KH domain-containing protein 2; RING finger protein 194; RING-type E3 ubiquitin transferase MEX3C; Rkhd2; RNA-binding E3 ubiquitin-protein ligase MEX3C; RNA-binding protein MEX3C; RNF194
Goat
Ammonium Sulfate Precipitation
RUO
240396, 51320
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q05A36, Q5U5Q3
MEX3C
Peptide with sequence C-TPRLSPTFPESIEH.
100 μg
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.