Learn More
RLBP1 Mouse anti-Human, Clone: OTI1B4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF503148
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a 36-kD water-soluble protein which carries 11-cis-retinaldehyde or 11-cis-retinal as physiologic ligands. It may be a functional component of the visual cycle. Mutations of this gene have been associated with severe rod-cone dystrophy, Bothnia dystrophy (nonsyndromic autosomal recessive retinitis pigmentosa) and retinitis punctata albescens.Specifications
| RLBP1 | |
| Monoclonal | |
| Unconjugated | |
| RLBP1 | |
| CRALBP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6017 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1B4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P12271 | |
| RLBP1 | |
| Full length human recombit protein of human RLBP1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.