missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RLBP1L1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-29652
This item is not returnable.
View return policy
Description
RLBP1L1 Polyclonal specifically detects RLBP1L1 in Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| RLBP1L1 | |
| Polyclonal | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:50, Immunocytochemistry 1:10 - 1:500 | |
| A6JFQ6 | |
| CLVS1 | |
| Synthetic peptide corresponding to amino acid residues from the C-terminal region conjugated to KLH. Accession # A6JFQ6 | |
| Affinity purified | |
| RUO | |
| Primary | |
| Specific for the ∼ 35 kDa clavesin 1/2 protein doublet in Western blots of rat brain lysate. Isoform-specific knock down in cultured hippocampal neurons indicates that the lower and upper bands are clavesin 1 and 2, respectively. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS and 50% Glycerol with No Preservative | |
| Cellular retinaldehyde-binding protein-like, clavesin 1, clavesin-1, CRALBPLC6orf212L, FLJ37248, MGC34646, retinaldehyde binding protein 1-like 1, Retinaldehyde-binding protein 1-like 1, RLBP1L1 | |
| Rabbit | |
| 35 kDa | |
| 0.1 mL | |
| Cellular Markers, Cellular Signaling, Neuronal Cell Markers | |
| 157807 | |
| Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction