missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RLF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563430
This item is not returnable.
View return policy
Description
Immunogen sequence: RYKCELNGCN IVFSDLGQLY HHEAQHFRDA SYTCNFLGCK KFYYSKIEYQ NHLSMHNVEN SNGDIKKSVK LEESATGEKQ DCINQPHLLN QTDKSHLPED LFCAESANSQ IDTETAENLK ENS Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 81%.
May be involved in transcriptional regulation.
Specifications
| RLF | |
| Polyclonal | |
| Unconjugated | |
| RLF | |
| rearranged L-myc fusion; rearranged L-myc fusion gene protein; rearranged L-myc fusion sequence; RLF; Zinc finger protein Rlf; Zn-15 related; ZN-15L; zn-15-related protein; ZNF292L | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6018 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13129 | |
| RLF | |
| Recombinant protein corresponding to Human RLF. Recombinant protein control fragment (Product #RP-89332). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction