missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RLTPR Antibody (EM-53), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62215MFV610
This item is not returnable.
View return policy
Description
RLTPR Monoclonal antibody specifically detects RLTPR in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Intracellular)
Specifications
| RLTPR | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 146206 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| EM-53 | |
| Flow Cytometry, Flow (Intracellular) | |
| CARMIL2, CARMIL2b, leucine rich repeat containing 16C, leucine-rich repeat-containing protein 16C, LRRC16C, RGD motif, leucine rich repeats, tropomodulin domain and proline-rich containing, RGD, leucine-rich repeat, tropomodulin and proline-rich containing protein, RGD, leucine-rich repeat, tropomodulin and proline-rich-containing protein | |
| Murine Rltpr (amino acids 1147-1397) | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction