missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RMND5A Antibody (OTI2C8), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73918FR
This item is not returnable.
View return policy
Description
RMND5A Monoclonal specifically detects RMND5A in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| RMND5A | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| RMND5A | |
| Recombinant protein fragment corresponding to amino acids 1-249 of human RMND5A (NP_073617) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI2C8 | |
| Western Blot | |
| C-terminal to LisH motif, 44 kDa, CTLH, FLJ12753, FLJ13910, FLJ21795, MGC78451, p44CTLH, protein RMD5 homolog A, required for meiotic nuclear division 5 homolog A (S. cerevisiae), RMD5 | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 64795.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction