missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RMND5A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17559-1-AP-20UL
This item is not returnable.
View return policy
Description
RMND5A is a protein coding gene.Specifications
| RMND5A | |
| Polyclonal | |
| Unconjugated | |
| RMND5A | |
| CTLH, p44CTLH, Protein RMD5 homolog A, RMD5, RMND5A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64795 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9H871 | |
| RMND5A | |
| RMND5A Fusion Protein Ag11751 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction