missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RNA polymerase I termination factor Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30191
This item is not returnable.
View return policy
Description
RNA polymerase I termination factor Polyclonal antibody specifically detects RNA polymerase I termination factor in Human samples. It is validated for Western Blot
Specifications
| RNA polymerase I termination factor | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7270 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RNA polymerase I termination factor, transcription termination factor 1, Transcription termination factor I, transcription termination factor, RNA polymerase I, TTF-1, TTF-I | |
| between 1 and 50 | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction