Learn More
RNA Polymerase II/POLR2A, p Ser2 Antibody (C.15200005), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59215
This item is not returnable.
View return policy
Description
RNA Polymerase II/POLR2A (p Ser2) Monoclonal antibody specifically detects RNA Polymerase II/POLR2A (p Ser2) in Human, C. elegans, Zebrafish samples. It is validated for Western Blot, ChIP assay, ELISA, Immunocytochemistry, Immunofluorescence, Knockdown.
Specifications
| RNA Polymerase II/POLR2A (p Ser2) | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| POLR2A | |
| The exact immunogen is propietary information. | |
| 50 μg | |
| Chromatin Research, Epigenetics, microRNA, Phospho Specific, Transcription Factors and Regulators | |
| 5430 | |
| Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, ChIP Assay | |
| C.15200005 | |
| Western Blot 1:1000, ELISA 1:3000, Immunocytochemistry/Immunofluorescence 1:500, Chromatin Immunoprecipitation (ChIP) 1-2 ug/IP, Chromatin Immunoprecipitation Sequencing 1 - 2 ug/ChIP, Knockdown Validated | |
| DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, C. elegans, Zebrafish | |
| Purified |