missing translation for 'onlineSavingsMsg'
Learn More

RNA Polymerase II/POLR2A Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 201.00 - € 520.00

Specifications

Antigen RNA Polymerase II/POLR2A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18396734
View Documents
Novus Biologicals
NBP3-16906-20UL
20 μg
€ 201.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18317194
View Documents
Novus Biologicals
NBP3-16906-100UL
100 μg
€ 520.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RNA Polymerase II/POLR2A Polyclonal antibody specifically detects RNA Polymerase II/POLR2A in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

RNA Polymerase II/POLR2A
Polyclonal
Purified
RUO
Human, Mouse
DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2
A synthetic phosphorylated peptide around S7 of RNA Polymerase II/POLR2A (NP_000928.1).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
Rabbit
Chromatin Research, Epigenetics, Phospho Specific, Transcription Factors and Regulators
PBS, 50% glycerol, pH7.3
5430
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.