missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNA polymerase sigma 70 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144478
This item is not returnable.
View return policy
Description
RNA polymerase sigma 70 Polyclonal Antibody for Western Blot, ELISA
RNA polymerase requires one of a family of sigma subunits, including sigma 70, for specific promoter recognition and initiation.
Specifications
| RNA polymerase sigma 70 | |
| Polyclonal | |
| Unconjugated | |
| rpoD | |
| RNA polymerase sigma factor RpoD (Sigma-70) rpoD alt b3067 JW3039 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 947567 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 3.43 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P00579 | |
| rpoD | |
| Recombinant Escherichia coli (strain K12) RPOD protein. | |
| 100 μL | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction