missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNase L Antibody (2E9), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-351B
This item is not returnable.
View return policy
Description
RNase L Monoclonal specifically detects RNase L in Human, Hamster, Primate, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RNase L | |
| Monoclonal | |
| Biotin | |
| Q05823 | |
| RNASEL | |
| Purified full-length recombinant human RNaseL produced in insect cells. [UniProt# Q05823] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, hamster and primate. This antibody does not react with mouse or rat. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Immunoassay, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 2000000000 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| 2-5A-dependent ribonuclease, 2-5A-dependent RNase, 2'-5'-oligoisoadenylate synthetase-dependent, EC 3.1.26.-, HPC1, interferon-induced 2-5A-dependent RNase, MGC104972, MGC133329, PRCA1, prostate cancer 1, Ribonuclease 4, Ribonuclease L, ribonuclease L (2'-5'-oligoisoadenylate synthetase-dependent), RNase L, RNS4DKFZp781D08126 | |
| Mouse | |
| 83 kDa | |
| 0.1 mL | |
| Chromatin Research, Cytokine Research, DNA replication Transcription Translation and Splicing, Immunology, Innate Immunity, Prostate Cancer, Protein Kinase | |
| 6041 | |
| Human, Hamster, Primate, Mouse (Negative), Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction